Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-3-hydroxy-3-methylbutanoic acid 98%

Supplier: AMBEED, INC
Warning

A314989-100MG A314989-250MG
77002-760EA 56 USD InStock
77002-760 77002-762
(R)-2-Amino-3-hydroxy-3-methylbutanoic acid 98%
(R)-2-Amino-3-hydroxy-3-methylbutanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
MDL Number: MFCD00145211
CAS Number: 2280-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty