Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LY2784544 ≥98%

Supplier: AMBEED, INC
Warning

A315267-10MG A315267-1MG A315267-5MG
76989-024EA 183 USD InStock
76989-024 76989-026 76989-028
LY2784544 ≥98%
LY2784544
Precaución: for Research Use Only


Formula: C₂₃H₂₅ClFN₇O
MW: 469.9 g/mol
MDL Number: MFCD20482143
CAS Number: 1229236-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty