Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-3-pyridineboronic acid pinacol ester 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A316389-10G A316389-1G A316389-250MG A316389-5G A316389-25G
76989-042EA 36 USD InStock
76989-042 76989-044 76989-046 76989-048 77683-941
4-Methyl-3-pyridineboronic acid pinacol ester 98%, Ambeed.Inc
4-Methyl-3-pyridineboronic acid pinacol ester
Precaución: For research use only.


Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD12923382
CAS Number: 1171891-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty