Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A316639-1000G A316639-100G A316639-10G A316639-25G A316639-500G A316639-5G
77002-790EA 110 USD InStock
77002-790 77002-792 77002-794 77002-796 77002-798 77002-800
2-Ethoxybenzoic acid 98%
2-Ethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 301
Melting Pt: 25
MDL Number: MFCD00002438
CAS Number: 134-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty