Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylamino)benzeneboronic acid, pinacol ester, N-BOC protected 96%

Supplier: AMBEED, INC
Warning

A317269-1G A317269-250MG A317269-5G
77015-220EA 43 USD InStock
77015-220 77015-222 77015-224
4-(Methylamino)benzeneboronic acid, pinacol ester, N-BOC protected 96%
4-(Methylamino)benzeneboronic acid, pinacol ester, N-BOC protected
Precaución: for Research Use Only


Formula: C₁₈H₂₈BNO₄
MW: 333.23 g/mol
MDL Number: MFCD03490498
CAS Number: 916587-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty