Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione 97%

Supplier: AMBEED, INC
Warning

A318950-100MG A318950-1G A318950-250MG A318950-5G
77032-734EA 14 USD InStock
77032-734 77032-736 77032-738 77032-740
4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione 97%
4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione
Precaución: for Research Use Only


Formula: C₁₁H₉F₃O₃
MDL Number: MFCD03419769
CAS Number: 57965-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty