Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester 98%

Supplier: AMBEED, INC
Warning

A319146-1G A319146-250MG A319146-5G
77032-742EA 12 USD InStock
77032-742 77032-744 77032-746
2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester 98%
2,3-Dihydro-1,4-benzodioxin-6-carboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₄
MDL Number: MFCD01544907
CAS Number: 20197-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty