Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-methyl-1,3-thiazole 97%

Supplier: AMBEED, INC
Warning

A319919-1G A319919-25G A319919-5G A319919-10G
77032-758EA 16 USD InStock
77032-758 77032-760 77032-762 77668-424
5-Bromo-4-methyl-1,3-thiazole 97%
5-Bromo-4-methyl-1,3-thiazole
Precaución: for Research Use Only


Formula: C₄H₄BrNS
MDL Number: MFCD12026322
CAS Number: 111600-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty