Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]furan-2-boronic acid pinacol ester 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A320167-250MG A320167-1G
76989-114EA 16 USD InStock
76989-114 77683-953
Benzo[b]furan-2-boronic acid pinacol ester 98%, Ambeed.Inc
Benzo[b]furan-2-boronic acid pinacol ester
Precaución: For research use only.


Formula: C₁₄H₁₇BO₃
MW: 244.094 g/mol
MDL Number: MFCD11870258
CAS Number: 402503-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty