Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%

Supplier: AMBEED, INC
Warning

A321520-100G A321520-25G A321520-500G A321520-5G
77002-862EA 51 USD InStock
77002-862 77002-864 77002-866 77002-868
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Precaución: for Research Use Only


Formula: C₁₀H₈F₆O
MDL Number: MFCD03093010
CAS Number: 127852-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty