Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) 98%

Supplier: AMBEED, INC
Warning

A326464-100G A326464-10G A326464-10MG A326464-1G A326464-50MG A326464-25G
77032-810EA 213 USD InStock
77032-810 77032-812 77032-814 77032-816 77032-818 77568-656
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) 98%
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Precaución: For research use only.


Formula: C₁₈H₁₈O₂
MDL Number: MFCD00016658
CAS Number: 528-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty