Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trimethoxybenzonitrile 95%

Supplier: AMBEED, INC
Warning

A326852-10G A326852-1G A326852-25G A326852-5G
77002-964EA 54 USD InStock
77002-964 77002-966 77002-968 77002-970
3,4,5-Trimethoxybenzonitrile 95%
3,4,5-Trimethoxybenzonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₃
MW: 193.2
MDL Number: MFCD00001803
CAS Number: 1885-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty