Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetradecyl pyridine-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A328129-1G A328129-250MG A328129-25G A328129-5G
77032-846EA 25 USD InStock
77032-846 77032-848 77032-850 77032-852
Tetradecyl pyridine-3-carboxylate 97%
Tetradecyl pyridine-3-carboxylate
Precaución: for Research Use Only


Formula: C₂₀H₃₃NO₂
MDL Number: MFCD16620608
CAS Number: 273203-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty