Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium [2,2-biquinoline]-4,4-dicarboxylate trihydrate 98%

Supplier: AMBEED, INC
Warning

A329106-1G A329106-25G A329106-5G
76989-242EA 6 USD InStock
76989-242 76989-244 76989-246
Potassium [2,2-biquinoline]-4,4-dicarboxylate trihydrate 98%
Potassium [2,2-biquinoline]-4,4-dicarboxylate trihydrate
Precaución: for Research Use Only


Formula: C₂₀H₁₆K₂N₂O₇
MDL Number: MFCD00191791
CAS Number: 207124-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty