Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyrimidin-2-ylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A332028-100MG A332028-1G A332028-250MG
77015-424EA 14 USD InStock
77015-424 77015-426 77015-428
4-Pyrimidin-2-ylbenzoic acid 98%
4-Pyrimidin-2-ylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₈N₂O₂
MDL Number: MFCD01318668
CAS Number: 199678-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty