Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Bis(diphenylphosphino)pyridine 98%

Supplier: AMBEED, INC
Warning

A332369-100MG A332369-250MG
76989-284EA 35 USD InStock
76989-284 76989-286
2,6-Bis(diphenylphosphino)pyridine 98%
2,6-Bis(diphenylphosphino)pyridine
Precaución: for Research Use Only


Formula: C₂₉H₂₃NP₂
MW: 447.45 g/mol
MDL Number: MFCD04039760
CAS Number: 64741-27-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty