Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N5,N5-Dimethylpyridine-2,5-diamine 98%

Supplier: AMBEED, INC
Danger

A332903-1G A332903-250MG
77015-442EA 66 USD InStock
77015-442 77015-444
N5,N5-Dimethylpyridine-2,5-diamine 98%
N5,N5-Dimethylpyridine-2,5-diamine
Precaución: for Research Use Only


Formula: C₇H₁₁N₃
MDL Number: MFCD10009551
CAS Number: 39856-52-5
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty