Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-4-methylphthalazine 98%

Supplier: AMBEED, INC
Warning

A333315-100MG A333315-1G A333315-250MG A333315-5G
77032-916EA 20 USD InStock
77032-916 77032-918 77032-920 77032-922
1-Chloro-4-methylphthalazine 98%
1-Chloro-4-methylphthalazine
Precaución: for Research Use Only


Formula: C₉H₇ClN₂
MW: 178.6183 g/mol
MDL Number: MFCD00631939
CAS Number: 19064-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty