Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine 95%

Supplier: AMBEED, INC
Warning

A334229-10G A334229-1G A334229-250MG A334229-5G
77032-970EA 72 USD InStock
77032-970 77032-972 77032-974 77032-976
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine 95%
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₅H₃BrF₃N₃
MW: 241.999 g/mol
MDL Number: MFCD09261254
CAS Number: 935534-47-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty