Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-(-)-1-Amino-2-indanol 98%

Supplier: AMBEED, INC
Warning

A335907-100MG A335907-250MG A335907-25G A335907-5G
77003-058EA 12 USD InStock
77003-058 77003-060 77003-062 77672-718
(1R,2R)-(-)-1-Amino-2-indanol 98%
(1R,2R)-(-)-1-Amino-2-indanol
Precaución: for Research Use Only


Formula: C₉H₁₁NO
MDL Number: MFCD08275447
CAS Number: 163061-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty