Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Fluorophenyl)-N'-(4-hydroxyphenyl)cyclopropane-1,1-dicarboxamide 95%

Supplier: AMBEED, INC
Warning

A336996-100MG A336996-1G A336996-250MG A336996-25G A336996-5G
76989-358EA 11 USD InStock
76989-358 76989-360 76989-362 76989-364 76989-366
N-(4-Fluorophenyl)-N'-(4-hydroxyphenyl)cyclopropane-1,1-dicarboxamide 95%
N-(4-Fluorophenyl)-N'-(4-hydroxyphenyl)cyclopropane-1,1-dicarboxamide
Precaución: for Research Use Only


Formula: C₁₇H₁₅FN₂O₃
MDL Number: MFCD22045581
CAS Number: 849217-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty