Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperidine-4-carbothioamide 95%

Supplier: AMBEED, INC
Warning

A341105-1G A341105-250MG A341105-25G A341105-5G
77015-576EA 46 USD InStock
77015-576 77015-578 77015-580 77015-582
Piperidine-4-carbothioamide 95%
Piperidine-4-carbothioamide
Precaución: for Research Use Only


Formula: C₆H₁₂N₂S
MW: 144.23 g/mol
MDL Number: MFCD09055273
CAS Number: 112401-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty