Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-chloronaphthalene 95%

Supplier: AMBEED, INC
Warning

A343525-100MG A343525-1G A343525-250MG A343525-50MG
76989-480EA 30 USD InStock
76989-480 76989-482 76989-484 76989-486
2-Bromo-6-chloronaphthalene 95%
2-Bromo-6-chloronaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆BrCl
MDL Number: MFCD17012579
CAS Number: 870822-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty