Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Bromophenyl)naphthalene 95%

Supplier: AMBEED, INC
Warning

A344300-100G A344300-10G A344300-1G A344300-25G A344300-5G
76989-494EA 355 USD InStock
76989-494 76989-496 76989-498 76989-500 76989-502
2-(3-Bromophenyl)naphthalene 95%
2-(3-Bromophenyl)naphthalene
Precaución: for Research Use Only


Formula: C₁₆H₁₁Br
MDL Number: MFCD16658911
CAS Number: 667940-23-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty