Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl tetrahydropyran-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A344762-10G A344762-1G A344762-25G
77003-190EA 17 USD InStock
77003-190 77003-192 77572-004
Methyl tetrahydropyran-4-carboxylate 98%
Methyl tetrahydropyran-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₁₂O₃
MDL Number: MFCD01075170
CAS Number: 110238-91-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty