Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxycarbonyl-D-phenylglycine 97%

Supplier: AMBEED, INC
Warning

A345130-100G A345130-25G A345130-5G A345130-500G
77003-196EA 43 USD InStock
77003-196 77003-198 77003-200 77672-754
N-Methoxycarbonyl-D-phenylglycine 97%
N-Methoxycarbonyl-D-phenylglycine
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD12796004
CAS Number: 50890-96-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty