Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloropyridine-4-carboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A345408-10G A345408-1G A345408-250MG
77003-202EA 460 USD InStock
77003-202 77003-204 77003-206
2,6-Dichloropyridine-4-carboxaldehyde 98%
2,6-Dichloropyridine-4-carboxaldehyde
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO
MW: 176 g/mol
MDL Number: MFCD00124816
CAS Number: 113293-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty