Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A347729-1G A347729-250MG
76989-560EA 16 USD InStock
76989-560 76989-562
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride 95%
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₇Cl₂N₃
MDL Number: MFCD11506491
CAS Number: 1052538-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty