Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-((3-Chloro-4-(3,3,3-trifluoro-2-hydroxy-2-methylpropanamido)phenyl)sulfonyl)-N,N-dimethylbenzamide 98%

Supplier: AMBEED, INC
Warning

A349552-10MG A349552-1MG A349552-50MG A349552-5MG
76989-602EA 210 USD InStock
76989-602 76989-604 76989-606 76989-608
(R)-4-((3-Chloro-4-(3,3,3-trifluoro-2-hydroxy-2-methylpropanamido)phenyl)sulfonyl)-N,N-dimethylbenzamide 98%
(R)-4-((3-Chloro-4-(3,3,3-trifluoro-2-hydroxy-2-methylpropanamido)phenyl)sulfonyl)-N,N-dimethylbenzamide
Precaución: for Research Use Only


Formula: C₁₉H₁₈ClF₃N₂O₅S
MDL Number: MFCD25976792
CAS Number: 252017-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty