Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-methyl-6-oxo-1,6-dihydropyrimidin-5-yl)-N,N-dimethylethanethioamide 98%

Supplier: AMBEED, INC
Danger

A349742-100MG A349742-10MG A349742-1MG A349742-25MG A349742-50MG A349742-5MG
76989-610EA 19 USD InStock
76989-610 76989-612 76989-614 76989-616 76989-618 76989-620
2-(1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-methyl-6-oxo-1,6-dihydropyrimidin-5-yl)-N,N-dimethylethanethioamide 98%
2-(1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-methyl-6-oxo-1,6-dihydropyrimidin-5-yl)-N,N-dimethylethanethioamide
Precaución: for Research Use Only


Formula: C₂₇H₃₁N₇OS
MDL Number: MFCD13194795
CAS Number: 247257-48-3
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty