Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butoxy-2,6-difluorophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A353625-1G A353625-250MG A353625-5G
77015-742EA 59 USD InStock
77015-742 77015-744 77015-746
3-Butoxy-2,6-difluorophenylboronic acid 98%
3-Butoxy-2,6-difluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃BF₂O₃
MDL Number: MFCD07369754
CAS Number: 849062-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty