Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methoxy-5-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A354163-100MG A354163-1G A354163-250MG
76989-696EA 23 USD InStock
76989-696 76989-698 76989-700
3-Chloro-2-methoxy-5-methylpyridine 98%
3-Chloro-2-methoxy-5-methylpyridine
Precaución: for Research Use Only


Formula: C₇H₈ClNO
MW: 157.60 g/mol
MDL Number: MFCD16610597
CAS Number: 1227593-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty