Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromobut-1-yn-1-yl)trimethylsilane 95%

Supplier: AMBEED, INC
Danger

A355194-1G A355194-250MG
76989-730EA 18 USD InStock
76989-730 76989-732
(4-Bromobut-1-yn-1-yl)trimethylsilane 95%
(4-Bromobut-1-yn-1-yl)trimethylsilane


Formula: C₇H₁₃BrSi
MDL Number: MFCD24458737
CAS Number: 69361-41-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty