Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pyridin-2-ylpropan-1-one 97%

Supplier: AMBEED, INC
Warning

A357054-10G A357054-1G A357054-250MG A357054-25G
76989-758EA 439 USD InStock
76989-758 76989-760 76989-762 76989-764
1-Pyridin-2-ylpropan-1-one 97%
1-Pyridin-2-ylpropan-1-one
Precaución: for Research Use Only


Formula: C₈H₉NO
MDL Number: MFCD00023486
CAS Number: 3238-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty