Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrimidine-2-carbothioamide 98%

Supplier: AMBEED, INC
Warning

A358147-100G A358147-1G A358147-25G A358147-5G
77015-810EA 468 USD InStock
77015-810 77015-812 77015-814 77015-816
Pyrimidine-2-carbothioamide 98%
Pyrimidine-2-carbothioamide
Precaución: for Research Use Only


Formula: C₅H₅N₃S
MW: 139.18 g/mol
MDL Number: MFCD12093703
CAS Number: 4537-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty