Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl fumaroyl chloride 98%

Supplier: AMBEED, INC
Danger

A358259-1G A358259-250MG A358259-5G
77015-824EA 8.49 USD InStock
77015-824 77015-826 77015-828 77015-830
Ethyl fumaroyl chloride 98%
Ethyl fumaroyl chloride
Precaución: for Research Use Only


Formula: C₆H₇ClO₃
MW: 162.57 g/mol
MDL Number: MFCD03002066
CAS Number: 26367-48-6
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty