Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethylpyrazole-5-carbohydrazide ≥95%

Supplier: AMBEED, INC
Warning

A365714-10G A365714-1G A365714-250MG
77033-224EA 351 USD InStock
77033-224 77033-226 77033-228
1,3-Dimethylpyrazole-5-carbohydrazide ≥95%
1,3-Dimethylpyrazole-5-carbohydrazide
Precaución: for Research Use Only


Formula: C₆H₁₀N₄O
MDL Number: MFCD02253833
CAS Number: 89187-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty