Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-7-chloroquinoline 95%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A369002-100MG A369002-1G A369002-5G A369002-250MG
77015-950EA 13 USD InStock
77015-950 77015-952 77015-954 77684-070
4-Amino-7-chloroquinoline 95%, Ambeed.Inc
4-Amino-7-chloroquinoline
Precaución: For research use only.


Formula: C₉H₇ClN₂
MDL Number: MFCD00828822
CAS Number: 1198-40-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty