Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Amino(phenyl)methyl)cyclopropanol 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A369253-1G A369253-250MG A369253-100MG
76989-860EA 421 USD InStock
76989-860 76989-862 77684-073
1-(Amino(phenyl)methyl)cyclopropanol 95%, Ambeed.Inc
1-(Amino(phenyl)methyl)cyclopropanol
Precaución: For research use only.


Formula: C₁₀H₁₃NO
MW: 163.21 g/mol
MDL Number: MFCD22681561
CAS Number: 1391737-71-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty