Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl 97%

Supplier: AMBEED, INC
Warning

A369765-100MG A369765-1G A369765-250MG A369765-5G
77015-960EA 15 USD InStock
77015-960 77015-962 77015-964 77015-966
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl 97%
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂O
MDL Number: MFCD02916382
CAS Number: 81998-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty