Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 7-bromoisoquinoline-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A375605-100MG A375605-1G
77033-346EA 63 USD InStock
77033-346 77033-348
Ethyl 7-bromoisoquinoline-3-carboxylate 98%
Ethyl 7-bromoisoquinoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₀BrNO₂
MDL Number: MFCD13195308
CAS Number: 660830-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty