Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide 95%

Supplier: AMBEED, INC
Warning

A375650-1G A375650-250MG A375650-5G
77003-572EA 15 USD InStock
77003-572 77003-574 77003-576
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide 95%
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
Precaución: for Research Use Only


Formula: C₈H₁₂N₄O₃
MW: 212.21 g/mol
MDL Number: MFCD02253196
CAS Number: 139756-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty