Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenothiazine-10-carbonyl chloride 95%

Supplier: AMBEED, INC
Danger

A376024-1G A376024-25G
77003-578EA 13 USD InStock
77003-578 77003-580
Phenothiazine-10-carbonyl chloride 95%
Phenothiazine-10-carbonyl chloride
Precaución: for Research Use Only


Formula: C₁₃H₈ClNOS
MDL Number: MFCD00044216
CAS Number: 18956-87-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty