Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-[(methylamino)methyl]piperidine-1-carboxylate oxalate 95%

Supplier: AMBEED, INC
Danger

A378914-100MG A378914-1G A378914-250MG A378914-5G
77033-392EA 14 USD InStock
77033-392 77033-394 77033-396 77668-754
tert-Butyl-3-[(methylamino)methyl]piperidine-1-carboxylate oxalate 95%
tert-Butyl-3-[(methylamino)methyl]piperidine-1-carboxylate oxalate
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₂
MW: 228.33 g/mol
MDL Number: MFCD13857453
CAS Number: 1017356-25-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty