Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pentyl-1H-indole 95%

Supplier: AMBEED, INC
Warning

A381648-100MG A381648-10G A381648-1G A381648-250MG A381648-5G
76989-986EA 50 USD InStock
76989-986 76989-988 76989-990 76989-992 76989-994
1-Pentyl-1H-indole 95%
1-Pentyl-1H-indole
Precaución: for Research Use Only


Formula: C₁₃H₁₇N
MW: 187.28 g/mol
MDL Number: MFCD00963300
CAS Number: 59529-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty