Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraphenylcyclopentadienone 95%

Supplier: AMBEED, INC
Warning

A386373-100G A386373-1G A386373-25G A386373-5G
77003-696EA 211 USD InStock
77003-696 77003-698 77003-700 77003-702
Tetraphenylcyclopentadienone 95%
Tetraphenylcyclopentadienone
Precaución: for Research Use Only


Formula: C₂₉H₂₀O
MDL Number: MFCD00001407
CAS Number: 479-33-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty