Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Aminophenyl)propionic acid 95%

Supplier: AMBEED, INC
Warning

A388789-1G A388789-25G A388789-5G
77003-734EA 17 USD InStock
77003-734 77003-736 77003-738
3-(3-Aminophenyl)propionic acid 95%
3-(3-Aminophenyl)propionic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MDL Number: MFCD00153889
CAS Number: 1664-54-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty