Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-{2-[3-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 98%

Supplier: AMBEED, INC
Warning

A396799-100MG A396799-1G A396799-250MG A396799-5G
77016-262EA 39 USD InStock
77016-262 77016-264 77016-266 77016-268
4-{2-[3-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 98%
4-{2-[3-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine
Precaución: for Research Use Only


Formula: C₁₈H₂₈BNO₄
MDL Number: MFCD09746214
CAS Number: 756520-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty