Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(cyclopropylcarbonyl)piperazine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A397113-1G A397113-250MG A397113-25G A397113-5G
77003-836EA 13 USD InStock
77003-836 77003-838 77003-840 77003-842
1-(cyclopropylcarbonyl)piperazine hydrochloride 97%
1-(cyclopropylcarbonyl)piperazine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅ClN₂O
MDL Number: MFCD03428576
CAS Number: 1021298-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty