Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Thiazolyl)ethanone 98%

Supplier: AMBEED, INC
Warning

A401670-10G A401670-1G A401670-250MG A401670-5G
77033-672EA 155 USD InStock
77033-672 77033-674 77033-676 77672-968
1-(4-Thiazolyl)ethanone 98%
1-(4-Thiazolyl)ethanone
Precaución: for Research Use Only


Formula: C₅H₅NOS
MDL Number: MFCD03550493
CAS Number: 38205-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty